Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003964.1 | NP_001003964.1 | 11 - 12 | 1153 | 77 | |
XM_006256250.2 | XP_006256312.1 | 10 - 11 | 1176 | 77 | ||
XM_006256251.2 | XP_006256313.1 | 2 - 3 | 548 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |