Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034032.1 | NP_001029204.1 | 1 - 2 | 111 | 92 | |
GenBank mRNA | AB062135.1 | - | 1 - 2 | 113 | 92 | |
BC100072.1 | - | 1 - 2 | 111 | 7325259 | 92 | |
FQ214972.1 | - | 1 - 2 | 124 | 92 | ||
FQ219445.1 | - | 1 - 2 | 124 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |