Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009972.1 | NP_001009972.1 | 4 - 5 | 982 | 81 | |
XM_006256125.2 | XP_006256187.1 | 4 - 5 | 1040 | 81 | ||
GenBank mRNA | AY529104.1 | - | 4 - 5 | 982 | 81 | |
BC105838.1 | - | 4 - 5 | 990 | 7452960 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |