Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240647.2 | XP_006240709.1 | 2 | 3395 | 90 | |
XM_008764964.1 | XP_008763186.1 | 2 | 1611 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |