Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343107.7 | XP_343108.4 | 5 - 6 | 2014 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |