Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040128.1 | NP_001035217.1 | 7 - 8 | 1006 | 123 | |
XM_006227834.1 | XP_006227896.1 | 8 - 9 | 1168 | 123 | ||
XM_006227837.2 | XP_006227899.1 | 8 - 9 | 1685 | 123 | ||
GenBank mRNA | FQ233028.1 | - | 7 - 8 | 555 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |