Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004216.1 | NP_001004216.1 | 4 - 5 | 337 | 134 | |
GenBank mRNA | AY573239.1 | - | 4 - 5 | 337 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |