Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107459.1 | NP_001100929.1 | 3 - 4 | 373 | 113 | |
XM_006227854.2 | XP_006227916.1 | 2 - 3 | 314 | 113 | ||
XM_006227855.2 | XP_006227917.1 | 2 - 3 | 335 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |