Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106757.2 | NP_001100227.1 | 5 - 6 | 769 | 90 | |
GenBank mRNA | BC158889.1 | - | 5 - 6 | 769 | 8375684 | 90 |
FQ212442.1 | - | 5 - 6 | 804 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |