Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031546.1 | NP_113734.1 | 2 - 3 | 263 | 112 | |
GenBank mRNA | AB037934.1 | - | 2 - 3 | 263 | 112 | |
BC078794.1 | - | 2 - 3 | 258 | 7126233 | 112 | |
D38467.1 | - | 2 - 3 | 257 | 112 | ||
FQ219778.1 | - | 2 - 3 | 266 | 112 | ||
X69021.1 | - | 2 - 3 | 252 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |