Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008383.1 | NP_001008384.1 | 9 - 10 | 1340 | 102 | |
XM_006255517.2 | XP_006255579.1 | 10 - 11 | 1361 | 102 | ||
XM_008772521.1 | XP_008770743.1 | 10 - 11 | 1558 | 102 | ||
GenBank mRNA | BC085826.1 | - | 9 - 10 | 1340 | 7131855 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |