Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107162.1 | NP_001100632.1 | 4 - 5 | 497 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |