Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025722.2 | NP_001020893.2 | 3 - 4 | 797 | 105 | |
NM_001109936.1 | NP_001103406.1 | 4 - 5 | 346 | 105 | ||
NM_001109937.1 | NP_001103407.1 | 4 - 5 | 307 | 105 | ||
GenBank mRNA | BC098804.1 | - | 3 - 4 | 797 | 7376321 | 105 |
DN932869.1 | - | 4 - 5 | 342 | 105 | ||
DN935585.1 | - | 4 - 5 | 366 | 105 | ||
DY320517.1 | - | 4 - 5 | 347 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |