Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191557.1 | NP_001178486.1 | 3 - 4 | 487 | 95 | |
XM_006255291.2 | XP_006255353.1 | 4 - 5 | 2037 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |