Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106191.1 | NP_001099661.1 | 10 - 11 | 1414 | 60 | |
XM_006255751.2 | XP_006255813.1 | 10 - 11 | 1460 | 60 | ||
XM_006255752.2 | XP_006255814.1 | 10 - 11 | 1450 | 60 | ||
XM_006255753.2 | XP_006255815.1 | 9 - 10 | 1342 | 60 | ||
XM_006255754.2 | XP_006255816.1 | 9 - 10 | 1333 | 60 | ||
XM_006255755.2 | XP_006255817.1 | 10 - 11 | 1459 | 60 | ||
XM_006255756.2 | XP_006255818.1 | 10 - 11 | 1450 | 60 | ||
XM_006255757.2 | XP_006255819.1 | 9 - 10 | 1332 | 60 | ||
XM_008772601.1 | XP_008770823.1 | 10 - 11 | 1681 | 60 | ||
XM_008772602.1 | XP_008770824.1 | 10 - 11 | 1460 | 60 | ||
GenBank mRNA | BC160844.1 | - | 10 - 11 | 1441 | 8361869 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |