Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255650.2 | XP_006255712.1 | 5 - 6 | 1042 | 71 | |
XM_006255653.2 | XP_006255715.1 | 4 - 5 | 940 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |