Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012668.2 | NP_036800.1 | 5 - 6 | 607 | 139 | |
GenBank mRNA | BC089813.1 | - | 5 - 6 | 607 | 7372826 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |