Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022962.1 | NP_075251.1 | 16 - 17 | 3261 | 102 | |
GenBank mRNA | AF081144.1 | - | 15 - 16 | 3246 | 102 | |
AF081145.1 | - | 15 - 16 | 3246 | 102 | ||
AF081146.1 | - | 16 - 17 | 3261 | 102 | ||
AF081147.1 | - | 16 - 17 | 3261 | 102 | ||
AF111099.1 | - | 17 - 18 | 3587 | 102 | ||
U72487.1 | - | 15 - 16 | 3063 | 102 | ||
U78105.1 | - | 15 - 16 | 3388 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |