Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024237.1 | NP_001019408.1 | 3 - 4 | 392 | 102 | |
GenBank mRNA | BC094515.1 | - | 3 - 4 | 392 | 7314191 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |