Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107430.1 | NP_001100900.1 | 4 - 5 | 654 | 117 | |
XM_006255596.2 | XP_006255658.2 | 6 - 7 | 909 | 117 | ||
XM_008772514.1 | XP_008770736.1 | 5 - 6 | 825 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |