Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191700.1 | NP_001178629.1 | 2 - 3 | 464 | 117 | |
XM_006255402.2 | XP_006255464.1 | 2 - 3 | 800 | 117 | ||
XM_008772493.1 | XP_008770715.1 | 2 - 3 | 976 | 117 | ||
XM_008772494.1 | XP_008770716.1 | 2 - 3 | 532 | 117 | ||
XM_008772495.1 | XP_008770717.1 | 3 - 4 | 580 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |