Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004207.1 | NP_001004207.1 | 4 - 5 | 665 | 110 | |
GenBank mRNA | BC079188.1 | - | 4 - 5 | 665 | 7131199 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |