Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037193.1 | NP_001032270.1 | 1 - 2 | 132 | 75 | |
XM_008772496.1 | XP_008770718.1 | 1 - 2 | 186 | 75 | ||
GenBank mRNA | BC099222.1 | - | 1 - 2 | 132 | 7388035 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |