Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108456.3 | NP_001101926.3 | 11 - 12 | 1429 | 86 | |
GenBank mRNA | BC168848.1 | - | 11 - 12 | 1322 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |