Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109132.1 | NP_001102602.1 | 1 - 2 | 491 | 87 | |
XM_006255598.2 | XP_006255660.1 | 1 - 2 | 517 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |