Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191558.1 | NP_001178487.1 | 13 - 14 | 1361 | 59 | |
XM_008772512.1 | XP_008770734.1 | 14 - 15 | 1472 | 59 | ||
XM_008772513.1 | XP_008770735.1 | 13 - 14 | 1422 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |