Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191558.1 | NP_001178487.1 | 9 - 10 | 825 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |