Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009640.1 | NP_001009640.1 | 7 - 8 | 944 | 61 | |
XM_006255562.1 | XP_006255624.1 | 7 - 8 | 990 | 61 | ||
XM_006255563.2 | XP_006255625.1 | 7 - 8 | 1153 | 61 | ||
XM_008772488.1 | XP_008770710.1 | 7 - 8 | 943 | 61 | ||
GenBank mRNA | BC088461.1 | - | 7 - 8 | 944 | 7127580 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |