Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242627.1 | NP_001229556.1 | 7 - 8 | 1018 | 94 | |
XM_008772646.1 | XP_008770868.1 | 6 - 7 | 4486 | 94 | ||
XM_008772647.1 | XP_008770869.1 | 6 - 7 | 4486 | 94 | ||
XM_008772649.1 | XP_008770871.1 | 5 - 6 | 960 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |