Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242627.1 | NP_001229556.1 | 8 - 9 | 1111 | 84 | |
XM_008772647.1 | XP_008770869.1 | 7 - 8 | 4579 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |