Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106183.1 | NP_001099653.1 | 2 - 3 | 375 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |