Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012164.1 | NP_001012164.1 | 9 - 10 | 946 | 85 | |
XM_006255278.2 | XP_006255340.1 | 9 - 10 | 1613 | 85 | ||
XM_006255279.2 | XP_006255341.1 | 8 - 9 | 1478 | 85 | ||
XM_006255280.1 | XP_006255342.1 | 10 - 11 | 1195 | 85 | ||
XM_006255281.2 | XP_006255343.1 | 7 - 8 | 1331 | 85 | ||
GenBank mRNA | BC083857.1 | - | 9 - 10 | 946 | 7125718 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Probe spans exons |