Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108446.1 | NP_001101916.1 | 5 - 6 | 535 | 141 | |
XM_006255407.2 | XP_006255469.1 | 5 - 6 | 552 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |