Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108446.1 | NP_001101916.1 | 7 - 8 | 741 | 98 | |
XM_006255407.2 | XP_006255469.1 | 7 - 8 | 758 | 98 | ||
GenBank mRNA | AY724536.1 | - | 7 - 8 | 690 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |