Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106669.1 | NP_001100139.1 | 3 - 4 | 744 | 79 | |
XM_006238475.2 | XP_006238537.1 | 3 - 4 | 767 | 79 | ||
XM_006238476.2 | XP_006238538.1 | 3 - 4 | 758 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |