Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106669.1 | NP_001100139.1 | 6 - 7 | 1150 | 89 | |
XM_006238475.2 | XP_006238537.1 | 6 - 7 | 1173 | 89 | ||
XM_006238476.2 | XP_006238538.1 | 6 - 7 | 1164 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |