Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238423.2 | XP_006238485.1 | 10 - 11 | 1443 | 66 | |
XM_006238424.2 | XP_006238486.1 | 10 - 11 | 1449 | 66 | ||
XM_008763915.1 | XP_008762137.1 | 10 - 11 | 1443 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |