Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007632.2 | NP_001007633.1 | 1 - 2 | 66 | 63 | |
GenBank mRNA | CK357599.1 | - | 1 - 2 | 66 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |