Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033656.1 | NP_001028828.1 | 8 - 9 | 1734 | 63 | |
XM_006256497.1 | XP_006256559.1 | 8 - 9 | 1606 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |