Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014033.1 | NP_001014055.1 | 4 - 5 | 639 | 131 | |
XM_006256415.2 | XP_006256477.1 | 5 - 6 | 864 | 131 | ||
XM_006256417.2 | XP_006256479.1 | 4 - 5 | 857 | 131 | ||
XM_006256418.2 | XP_006256480.1 | 5 - 6 | 863 | 131 | ||
XM_008772886.1 | XP_008771108.1 | 4 - 5 | 1142 | 131 | ||
GenBank mRNA | BC087157.1 | - | 4 - 5 | 639 | 7133379 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |