Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020074.2 | NP_064459.1 | 2 - 3 | 245 | 118 | |
XM_006256483.2 | XP_006256545.1 | 5 - 6 | 625 | 118 | ||
XM_006256484.2 | XP_006256546.1 | 3 - 4 | 433 | 118 | ||
GenBank mRNA | BC088144.1 | - | 2 - 3 | 245 | 7384101 | 118 |
FQ220127.1 | - | 2 - 3 | 264 | 118 | ||
FQ227535.1 | - | 2 - 3 | 262 | 118 | ||
FQ231972.1 | - | 2 - 3 | 262 | 118 | ||
J03224.1 | - | 2 - 3 | 258 | 118 | ||
K02934.1 | - | 2 - 3 | 260 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |