Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106398.1 | NP_001099868.1 | 4 - 5 | 504 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |