Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191604.1 | NP_001178533.1 | 1698 | 66 | ||
XM_006256402.2 | XP_006256464.1 | 770 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |