Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191604.1 | NP_001178533.1 | 1344 | 78 | ||
XM_006256402.2 | XP_006256464.1 | 416 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |