Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191720.1 | NP_001178649.1 | 3 - 4 | 2794 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |