Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139483.1 | NP_001132955.1 | 1 - 2 | 206 | 119 | |
XM_006256452.2 | XP_006256514.1 | 2 - 3 | 320 | 119 | ||
XM_006256453.2 | XP_006256515.1 | 2 - 3 | 320 | 119 | ||
XM_006256454.2 | XP_006256516.1 | 2 - 3 | 263 | 119 | ||
XM_006256456.2 | XP_006256518.1 | 2 - 3 | 290 | 119 | ||
XM_006256459.2 | XP_006256521.1 | 2 - 3 | 320 | 119 | ||
XM_008772888.1 | XP_008771110.1 | 2 - 3 | 320 | 119 | ||
XM_008772889.1 | XP_008771111.1 | 2 - 3 | 278 | 119 | ||
XM_008772890.1 | XP_008771112.1 | 2 - 3 | 265 | 119 | ||
GenBank mRNA | AW521424.1 | - | 1 - 2 | 109 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |