Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 1 - 2 | 113 | 70 | |
GenBank mRNA | BC088442.1 | - | 1 - 2 | 113 | 7126325 | 70 |
FQ212547.1 | - | 1 - 2 | 112 | 70 | ||
FQ212814.1 | - | 1 - 2 | 62 | 70 | ||
FQ219838.1 | - | 1 - 2 | 79 | 70 | ||
FQ225424.1 | - | 1 - 2 | 84 | 70 | ||
FQ226083.1 | - | 1 - 2 | 59 | 70 | ||
FQ226422.1 | - | 1 - 2 | 62 | 70 | ||
FQ227142.1 | - | 1 - 2 | 62 | 70 | ||
FQ227488.1 | - | 1 - 2 | 111 | 70 | ||
FQ227830.1 | - | 1 - 2 | 62 | 70 | ||
FQ230874.1 | - | 1 - 2 | 97 | 70 | ||
FQ231828.1 | - | 1 - 2 | 64 | 70 | ||
FQ231874.1 | - | 1 - 2 | 97 | 70 | ||
FQ234230.1 | - | 1 - 2 | 64 | 70 | ||
FQ235071.1 | - | 1 - 2 | 84 | 70 | ||
FQ235201.1 | - | 1 - 2 | 79 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |