Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024743.1 | NP_001019914.1 | 7 - 8 | 1324 | 97 | |
XM_006251538.1 | XP_006251600.1 | 7 - 8 | 1276 | 97 | ||
GenBank mRNA | BC084711.1 | - | 2 - 3 | 431 | 97 | |
BC097369.1 | - | 7 - 8 | 1324 | 7443949 | 97 | |
FQ214781.1 | - | 2 - 3 | 507 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |