Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108366.1 | NP_001101836.1 | 5 - 6 | 425 | 149 | |
XM_008770432.1 | XP_008768654.1 | 5 - 6 | 448 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |