Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108367.1 | NP_001101837.1 | 10 - 11 | 1853 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |